siRNA Details
| Gene Symbol: FAM9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205849.3 | NP_995321.1 | 7, 8 | 853 | ||
| XM_011545464.2 | XP_011543766.1 | 14, 15 | 1426 | |||
| GenBank mRNA | AF494344.1 | - | 7, 8 | 852 | ||
| AK097501.1 | - | 6, 7 | 797 | |||
| AK128470.1 | - | 7, 8 | 3214 | |||
| BC120955.1 | - | 6, 7 | 552 | 40115245 | ||
| BC120956.1 | - | 6, 7 | 552 | 40115246 | ||

FAM9B
Genomic Map