siRNA Details
| Gene Symbol: SLC24A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205850.2 | NP_995322.1 | 5 | 613 | ||
| XM_017022079.1 | XP_016877568.1 | 4 | 1714 | |||
| XM_017022080.1 | XP_016877569.1 | 4 | 1934 | |||
| GenBank mRNA | AF348468.1 | - | 5 | 613 | ||
| BC113628.1 | - | 5 | 568 | 8322680 | ||
| BC113630.1 | - | 5 | 568 | 8322682 | ||
| BC143950.1 | - | 5 | 568 | 9052468 | ||
| DQ665306.1 | - | 5 | 561 | |||
| DQ665307.1 | - | 4 | 381 | |||

SLC24A5
Genomic Map