siRNA Details
| Gene Symbol: C6orf201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104464.1 | 3, 4 | 768 | |||
| XM_011514613.2 | XP_011512915.1 | 4, 5 | 760 | |||
| GenBank mRNA | AK301893.1 | - | 3, 4 | 581 | ||
| BC066986.1 | - | 3, 4 | 757 | 4827732 | ||

C6orf201
Genomic Map