siRNA Details
| Gene Symbol: MS4A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206893.3 | NP_996776.2 | 8 | 1514 | ||
| GenBank mRNA | AK122633.1 | - | 8 | 1510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MS4A10| Gene Symbol: MS4A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206893.3 | NP_996776.2 | 8 | 1514 | ||
| GenBank mRNA | AK122633.1 | - | 8 | 1510 | ||