siRNA Details
| Gene Symbol: SPAG17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206996.2 | NP_996879.1 | 3 | 359 | ||
| XM_006710426.3 | XP_006710489.1 | 3 | 359 | |||
| XM_006710427.3 | XP_006710490.1 | 3 | 359 | |||
| XM_011540934.1 | XP_011539236.1 | 3 | 359 | |||
| XM_011540935.2 | XP_011539237.1 | 1 | 60 | |||
| GenBank mRNA | AK091816.1 | - | 3 | 345 | ||
| AY555274.1 | - | 3 | 345 | |||
| BX648110.1 | - | 3 | 360 | |||

SPAG17
Genomic Map