siRNA Details
| Gene Symbol: FBXW12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159929.1 | NP_001153401.1 | 5 | 455 | ||
| NM_207102.2 | NP_996985.2 | 6 | 629 | |||
| XM_017006224.1 | XP_016861713.1 | 5 | 642 | |||
| GenBank mRNA | AY247969.1 | - | 6 | 629 | ||
| BC101303.1 | - | 5 | 802 | 40024696 | ||
| BC101305.1 | - | 5 | 399 | 40024702 | ||

FBXW12
Genomic Map