siRNA Details
| Gene Symbol: NPSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300933.1 | NP_001287862.1 | 7 | 962 | ||
| NM_001300934.1 | NP_001287863.1 | 5 | 797 | |||
| NM_001300935.1 | NP_001287864.1 | 7 | 995 | |||
| NM_207172.1 | NP_997055.1 | 7 | 949 | |||
| NM_207173.1 | NP_997056.1 | 7 | 949 | |||
| GenBank mRNA | AY310326.1 | - | 7 | 949 | ||
| AY310327.1 | - | 7 | 949 | |||
| AY310328.1 | - | 7 | 916 | |||
| AY310330.1 | - | 6 | 845 | |||
| AY310331.1 | - | 6 | 855 | |||
| AY310332.1 | - | 5 | 751 | |||
| AY956446.1 | - | 7 | 854 | |||
| AY956447.1 | - | 7 | 825 | |||
| AY956448.1 | - | 7 | 825 | |||
| BC132693.1 | - | 7 | 866 | 40146715 | ||
| BC132695.1 | - | 7 | 866 | 40146717 | ||
| BC144380.1 | - | 5 | 668 | 9052901 | ||
More Information
Set Membership: |
|

NPSR1
Genomic Map