siRNA Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207311.2 | NP_997194.2 | 2 | 604 | ||
| XM_005253991.4 | XP_005254048.1 | 2 | 629 | |||
| XM_006719694.3 | XP_006719757.1 | 2 | 631 | |||
| XM_006719696.3 | XP_006719759.1 | 2 | 630 | |||
| XM_006719697.3 | XP_006719760.1 | 2 | 631 | |||
| XM_011538999.2 | XP_011537301.1 | 2 | 633 | |||
| XM_011539001.2 | XP_011537303.1 | 2 | 642 | |||
| GenBank mRNA | AK129960.1 | - | 3 | 547 | ||
| U88834.1 | - | 2 | 569 | |||

BICDL1
Genomic Map