siRNA Details
Gene Symbol: TMEM132E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304438.1 | NP_001291367.1 | 9 | 3840 | ||
GenBank mRNA | BC018318.1 | - | 1 | 1266 | ||
BC020591.1 | - | 7 | 2496 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM132E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304438.1 | NP_001291367.1 | 9 | 3840 | ||
GenBank mRNA | BC018318.1 | - | 1 | 1266 | ||
BC020591.1 | - | 7 | 2496 |