siRNA Details
| Gene Symbol: TMEM207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207316.2 | NP_997199.1 | 5 | 923 | ||
| GenBank mRNA | AY358607.1 | - | 5 | 923 | ||
| BC071780.1 | - | 5 | 914 | 4607584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM207| Gene Symbol: TMEM207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207316.2 | NP_997199.1 | 5 | 923 | ||
| GenBank mRNA | AY358607.1 | - | 5 | 923 | ||
| BC071780.1 | - | 5 | 914 | 4607584 | ||