siRNA Details
Gene Symbol: LOC100505841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008912.1 | XP_016864401.1 | 2 | 732 |
Gene Symbol: ZNF474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207317.2 | NP_997200.1 | 2 | 765 | ||
GenBank mRNA | AK057483.1 | - | 2 | 759 | ||
AK290985.1 | - | 2 | 695 | |||
AY461732.1 | - | 2 | 758 | |||
BC133022.1 | - | 1 | 440 | 40147044 | ||
BC133026.1 | - | 1 | 440 | 40147048 |
