siRNA Details
| Gene Symbol: ZNF474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207317.2 | NP_997200.1 | 2 | 1572 | ||
| GenBank mRNA | AK057483.1 | - | 2 | 1565 | ||
| AK290985.1 | - | 2 | 1502 | |||
| AY461732.1 | - | 2 | 1565 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF474| Gene Symbol: ZNF474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207317.2 | NP_997200.1 | 2 | 1572 | ||
| GenBank mRNA | AK057483.1 | - | 2 | 1565 | ||
| AK290985.1 | - | 2 | 1502 | |||
| AY461732.1 | - | 2 | 1565 | |||