siRNA Details
| Gene Symbol: ACSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207321.2 | NP_997204.2 | 5 | 827 | ||
| GenBank mRNA | AK126134.1 | - | 5 | 817 | ||
| BC063654.1 | - | 2 | 216 | |||
| BC104210.1 | - | 4 | 711 | |||
| BC104211.2 | - | 4 | 711 | |||
| BC112988.1 | - | 5 | 792 | |||
| BC148184.1 | - | 4 | 711 | |||

ACSM6
Genomic Map