siRNA Details
| Gene Symbol: NIPAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207330.2 | NP_997213.1 | 2 | 160 | ||
| XM_017007784.1 | XP_016863273.1 | 2 | 117 | |||
| GenBank mRNA | AK095299.1 | - | 2 | 119 | ||
| BC067881.1 | - | 2 | 158 | 30345943 | ||
| CB960388.1 | - | 2 | 153 | |||
| CR749484.1 | - | 2 | 159 | |||

NIPAL1
Genomic Map