siRNA Details
| Gene Symbol: FAM43B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207334.2 | NP_997217.1 | 1 | 1788 | ||
| GenBank mRNA | AK126900.1 | - | 1 | 1788 | ||
| BC142610.1 | - | 1 | 1431 | 40147211 | ||
| BC142686.1 | - | 1 | 1679 | 40148591 | ||

FAM43B
Genomic Map