siRNA Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207335.2 | NP_997218.2 | 5 | 4949 | ||
| XM_005247167.1 | XP_005247224.1 | 6 | 5413 | |||
| XM_005247168.4 | XP_005247225.1 | 6 | 5491 | |||
| GenBank mRNA | AK128176.1 | - | 5 | 3811 | ||
| BC041365.1 | - | 2 | 3308 | |||
| BX537865.1 | - | 5 | 3988 | |||

KBTBD12
Genomic Map