siRNA Details
| Gene Symbol: ZP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207341.3 | NP_997224.2 | 5 | 992 | ||
| XM_011544852.2 | XP_011543154.1 | 2 | 508 | |||
| XM_011544853.2 | XP_011543155.1 | 3 | 371 | |||
| GenBank mRNA | BC067899.1 | - | 1 | 591 | 5298273 | |
| CR741030.1 | - | 1 | 262 | |||
| EU794684.1 | - | 5 | 992 | |||

ZP1
Genomic Map