siRNA Details
| Gene Symbol: RNF214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077239.1 | NP_001070707.1 | 3 | 557 | ||
| NM_207343.3 | NP_997226.2 | 3 | 619 | |||
| GenBank mRNA | AK298923.1 | - | 3 | 551 | ||
| BC146891.1 | - | 3 | 557 | 9056819 | ||
| BC146901.1 | - | 3 | 557 | 9056829 | ||
| CN395747.1 | - | 3 | 570 | |||

RNF214
Genomic Map