siRNA Details
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207345.3 | NP_997228.1 | 9 | 1657 | ||
GenBank mRNA | AY358265.1 | - | 9 | 1657 | ||
DB526089.1 | - | 1 | 345 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207345.3 | NP_997228.1 | 9 | 1657 | ||
GenBank mRNA | AY358265.1 | - | 9 | 1657 | ||
DB526089.1 | - | 1 | 345 |