siRNA Details
| Gene Symbol: SLC25A34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207348.2 | NP_997231.1 | 4 | 872 | ||
| XM_011541292.2 | XP_011539594.1 | 4 | 2098 | |||
| XM_011541293.1 | XP_011539595.1 | 4 | 2098 | |||
| GenBank mRNA | BC027998.1 | - | 4 | 872 | 5189944 | |
| CR749264.1 | - | 1 | 109 | |||

SLC25A34
Genomic Map