siRNA Details
| Gene Symbol: SLC25A34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207348.2 | NP_997231.1 | 5 | 1450 | ||
| XM_011541292.2 | XP_011539594.1 | 6 | 2750 | |||
| XM_017001083.1 | XP_016856572.1 | 4 | 2541 | |||
| GenBank mRNA | AK024244.1 | - | 1 | 171 | ||
| AL832282.1 | - | 1 | 525 | |||
| BC027998.1 | - | 5 | 1450 | 5189944 | ||
| CR749264.1 | - | 2 | 687 | |||

SLC25A34
Genomic Map