siRNA Details
| Gene Symbol: ANKRD13D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207354.2 | NP_997237.2 | 14 | 1747 | ||
| NR_030767.1 | 15 | 1830 | ||||
| XM_011544985.1 | XP_011543287.1 | 15 | 1837 | |||
| XM_011544986.1 | XP_011543288.1 | 16 | 1977 | |||
| XM_011544987.2 | XP_011543289.1 | 15 | 1811 | |||
| XM_011544988.2 | XP_011543290.1 | 10 | 1167 | |||
| GenBank mRNA | AK027313.1 | - | 7 | 1688 | ||
| AK057681.1 | - | 7 | 2361 | |||
| AK091614.1 | - | 2 | 2780 | |||
| AK096277.1 | - | 14 | 1660 | |||
| AK124721.1 | - | 15 | 1827 | |||
| AK126438.1 | - | 13 | 1744 | |||
| BC044239.1 | - | 4 | 718 | |||
| BC110419.1 | - | 14 | 1720 | 5210878 | ||
| BC121024.1 | - | 14 | 1621 | 40117420 | ||
| BC121025.2 | - | 14 | 1621 | 40117423 | ||

ANKRD13D
Genomic Map