siRNA Details
| Gene Symbol: LOC101927506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014131.1 | XP_016869620.1 | 2 | 1941 | ||
| XM_017014132.1 | XP_016869621.1 | 2 | 1941 | |||
| XM_017014133.1 | XP_016869622.1 | 2 | 1941 | |||
| XM_017014134.1 | XP_016869623.1 | 2 | 1941 | |||
| XM_017014135.1 | XP_016869624.1 | 2 | 1941 | |||
| XM_017014136.1 | XP_016869625.1 | 2 | 1941 | |||
| Gene Symbol: LOC100134822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137655.1 | - | 17 | 3006 | ||
| Gene Symbol: SEPT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207366.2 | NP_997249.2 | 10 | 1250 | ||
| XM_011515373.2 | XP_011513675.1 | 10 | 1234 | |||
| GenBank mRNA | AK126048.1 | - | 5 | 538 | ||
| AK301928.1 | - | 10 | 1213 | |||
| DB087561.1 | - | 5 | 538 | |||

LOC101927506
Genomic Map