siRNA Details
| Gene Symbol: SKIDA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207371.3 | NP_997254.3 | 4 | 3235 | ||
| XM_005252450.3 | XP_005252507.1 | 5 | 2344 | |||
| XM_011519480.2 | XP_011517782.1 | 5 | 2341 | |||
| XM_017016225.1 | XP_016871714.1 | 5 | 3135 | |||
| XM_017016226.1 | XP_016871715.1 | 5 | 3138 | |||
| GenBank mRNA | AB234866.1 | - | 2 | 836 | ||
| AK127130.1 | - | 2 | 270 | |||
| AK131456.1 | - | 5 | 2965 | |||

SKIDA1
Genomic Map