siRNA Details
| Gene Symbol: LINC00671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027254.1 | 4 | 1229 | |||
| GenBank mRNA | AK055784.1 | - | 4 | 1339 | ||
| BC122867.1 | - | 3 | 1059 | |||
| BC122868.1 | - | 4 | 1151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00671| Gene Symbol: LINC00671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027254.1 | 4 | 1229 | |||
| GenBank mRNA | AK055784.1 | - | 4 | 1339 | ||
| BC122867.1 | - | 3 | 1059 | |||
| BC122868.1 | - | 4 | 1151 | |||