siRNA Details
| Gene Symbol: RGS9BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207391.2 | NP_997274.2 | 1 | 2615 | ||
| GenBank mRNA | AF086461.1 | - | 1 | 488 | ||
| AK124499.1 | - | 1 | 2616 | |||
| AK127646.1 | - | 1 | 2172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGS9BP| Gene Symbol: RGS9BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207391.2 | NP_997274.2 | 1 | 2615 | ||
| GenBank mRNA | AF086461.1 | - | 1 | 488 | ||
| AK124499.1 | - | 1 | 2616 | |||
| AK127646.1 | - | 1 | 2172 | |||