siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128246.1 | - | 4 | 1505 | ||
| BC119780.1 | - | 3 | 993 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF207| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128246.1 | - | 4 | 1505 | ||
| BC119780.1 | - | 3 | 993 | |||