siRNA Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134656.1 | NP_001128128.1 | 4 | 1601 | ||
| NM_207404.3 | NP_997287.2 | 5 | 1689 | |||
| GenBank mRNA | AK127779.1 | - | 5 | 1689 | ||
| AK131235.1 | - | 1 | 1139 | |||
| BC128085.1 | - | 3 | 1314 | 40108370 | ||

ZNF662
Genomic Map