siRNA Details
| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207407.2 | NP_997290.2 | 2 | 123 | ||
| XM_011531935.1 | XP_011530237.1 | 2 | 113 | |||
| GenBank mRNA | AK122625.1 | - | 2 | 123 | ||
| DA441628.1 | - | 2 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11F| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207407.2 | NP_997290.2 | 2 | 123 | ||
| XM_011531935.1 | XP_011530237.1 | 2 | 113 | |||
| GenBank mRNA | AK122625.1 | - | 2 | 123 | ||
| DA441628.1 | - | 2 | 113 | |||