siRNA Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286550.1 | NP_001273479.1 | 4 | 488 | ||
| NM_207409.3 | NP_997292.2 | 3 | 409 | |||
| NR_104467.1 | 3 | 511 | ||||
| NR_104469.1 | 2 | 388 | ||||
| GenBank mRNA | AY358823.1 | - | 4 | 467 | ||
| CD513822.1 | - | 3 | 538 | |||

CLPSL2
Genomic Map