siRNA Details
| Gene Symbol: MROH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207414.2 | NP_997297.2 | 21 | 2818 | ||
| NR_102363.2 | 19 | 2478 | ||||
| NR_102364.2 | 20 | 2749 | ||||
| GenBank mRNA | AK125848.1 | - | 21 | 2818 | ||
| AK131461.1 | - | 20 | 2749 | |||
| BC144560.1 | - | 19 | 2454 | |||

MROH5
Genomic Map