siRNA Details
| Gene Symbol: MROH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207414.2 | NP_997297.2 | 24 | 3194 | ||
| NR_102363.2 | 22 | 2854 | ||||
| NR_102364.2 | 23 | 3125 | ||||
| GenBank mRNA | AK125848.1 | - | 24 | 3194 | ||
| AK131461.1 | - | 23 | 3125 | |||
| BC144560.1 | - | 22 | 2830 | |||

MROH5
Genomic Map