siRNA Details
| Gene Symbol: MROH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207414.2 | NP_997297.2 | 29, 30 | 3932 | ||
| NR_102363.2 | 27, 28 | 3592 | ||||
| NR_102364.2 | 26, 27 | 3583 | ||||
| GenBank mRNA | AK125848.1 | - | 29, 30 | 3932 | ||
| AK131461.1 | - | 26, 27 | 3583 | |||
| BC144560.1 | - | 27, 28 | 3568 | |||

MROH5
Genomic Map