siRNA Details
| Gene Symbol: CFAP77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282957.1 | NP_001269886.1 | 3 | 429 | ||
| NM_207417.2 | NP_997300.1 | 4 | 537 | |||
| XM_011518670.2 | XP_011516972.1 | 3 | 582 | |||
| XM_011518671.1 | XP_011516973.1 | 3 | 582 | |||
| XM_017014711.1 | XP_016870200.1 | 3 | 582 | |||
| GenBank mRNA | AK128819.1 | - | 4 | 509 | ||
| BC118593.1 | - | 3 | 397 | 40108008 | ||
| BC124555.1 | - | 3 | 397 | 40108012 | ||
| HY017665.1 | - | 3 | 430 | |||

CFAP77
Genomic Map