siRNA Details
| Gene Symbol: CFAP77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282957.1 | NP_001269886.1 | 6 | 1313 | ||
| NM_207417.2 | NP_997300.1 | 7 | 1421 | |||
| GenBank mRNA | AK128819.1 | - | 7 | 1393 | ||
| BC118593.1 | - | 6 | 1281 | 40108008 | ||
| BC124555.1 | - | 6 | 1281 | 40108012 | ||

CFAP77
Genomic Map