siRNA Details
| Gene Symbol: C12orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207435.2 | NP_997318.1 | 5 | 1219 | ||
| XM_017019297.1 | XP_016874786.1 | 4 | 887 | |||
| GenBank mRNA | AK021447.1 | - | 3 | 814 | ||
| AK097461.1 | - | 5 | 1219 | |||
| BC041968.1 | - | 2 | 715 | |||
| BC133010.1 | - | 4 | 1002 | 40147032 | ||
| BC133012.1 | - | 4 | 1003 | 40147034 | ||
| BC144372.1 | - | 4 | 1093 | |||

C12orf76
Genomic Map