siRNA Details
| Gene Symbol: SMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004595.4 | NP_004586.2 | 3 | 504 | ||
| XM_005274582.2 | XP_005274639.1 | 3 | 570 | |||
| XM_011545568.2 | XP_011543870.1 | 3 | 364 | |||
| XM_017029753.1 | XP_016885242.1 | 3 | 344 | |||
| XM_017029754.1 | XP_016885243.1 | 3 | 311 | |||
| XM_017029755.1 | XP_016885244.1 | 3 | 402 | |||
| GenBank mRNA | AD001528.1 | - | 3 | 353 | ||
| AK313834.1 | - | 3 | 336 | |||
| BC009898.2 | - | 3 | 327 | 3506022 | ||
| BC085621.1 | - | 3 | 296 | 5540463 | ||
| Z49099.1 | - | 3 | 280 | |||

SMS
Genomic Map