siRNA Details
Gene Symbol: C18orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272093.2 | NP_001259022.2 | 2 | 1394 | ||
GenBank mRNA | AK126293.1 | - | 2 | 1394 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C18orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272093.2 | NP_001259022.2 | 2 | 1394 | ||
GenBank mRNA | AK126293.1 | - | 2 | 1394 |