siRNA Details
| Gene Symbol: FAM177B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136691.1 | 5 | 724 | |||
| XM_017001283.1 | XP_016856772.1 | 4 | 498 | |||
| GenBank mRNA | AK125494.1 | - | 5 | 724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM177B| Gene Symbol: FAM177B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136691.1 | 5 | 724 | |||
| XM_017001283.1 | XP_016856772.1 | 4 | 498 | |||
| GenBank mRNA | AK125494.1 | - | 5 | 724 | ||