siRNA Details
| Gene Symbol: FSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173651.3 | NP_775922.3 | 17 | 19593 | ||
| GenBank mRNA | AK126036.1 | - | 1 | 1910 | ||
| AK126051.1 | - | 1 | 3097 | |||
| BX647691.1 | - | 2 | 2688 | |||
| BX648733.1 | - | 1 | 3866 | |||

FSIP2
Genomic Map