siRNA Details
| Gene Symbol: RAB44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257357.1 | NP_001244286.1 | 14 | 3267 | ||
| GenBank mRNA | AK125083.1 | - | 6 | 821 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB44| Gene Symbol: RAB44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257357.1 | NP_001244286.1 | 14 | 3267 | ||
| GenBank mRNA | AK125083.1 | - | 6 | 821 | ||