siRNA Details
| Gene Symbol: TSPAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004615.3 | NP_004606.2 | 4 | 428 | ||
| GenBank mRNA | AB062057.1 | - | 4 | 429 | ||
| AB102665.1 | - | 4 | 344 | |||
| AK293178.1 | - | 5 | 413 | |||
| AK293534.1 | - | 6 | 582 | |||
| AK293552.1 | - | 5 | 493 | |||
| AK312343.1 | - | 4 | 362 | |||
| BC018036.1 | - | 4 | 387 | 4801388 | ||
| D10653.1 | - | 4 | 368 | |||
| D29808.1 | - | 4 | 354 | |||
| L10373.1 | - | 4 | 412 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019634, AK004776, BC026516, BC058603, BY238972, and others.

TSPAN7
Genomic Map