siRNA Details
| Gene Symbol: C7orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201450.1 | NP_001188379.1 | 5 | 1707 | ||
| NM_001201451.1 | NP_001188380.1 | 4 | 1677 | |||
| GenBank mRNA | AK124274.1 | - | 5 | 1707 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf76| Gene Symbol: C7orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201450.1 | NP_001188379.1 | 5 | 1707 | ||
| NM_001201451.1 | NP_001188380.1 | 4 | 1677 | |||
| GenBank mRNA | AK124274.1 | - | 5 | 1707 | ||