siRNA Details
| Gene Symbol: C11orf87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207645.3 | NP_997528.2 | 2 | 1395 | ||
| XM_011542817.2 | XP_011541119.1 | 2 | 1499 | |||
| XM_011542818.2 | XP_011541120.1 | 2 | 1377 | |||
| GenBank mRNA | AB096240.1 | - | 1 | 503 | ||
| AK303693.1 | - | 2 | 1395 | |||
| BC035798.1 | - | 2 | 1315 | 5735100 | ||
| BC068577.1 | - | 2 | 1368 | 4818568 | ||

C11orf87
Genomic Map