siRNA Details
| Gene Symbol: Phxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119390.1 | - | 635 | 8734139 | ||
| BC119392.1 | - | 635 | 8734141 | |||
| X12808.1 | - | 641 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phxr2| Gene Symbol: Phxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119390.1 | - | 635 | 8734139 | ||
| BC119392.1 | - | 635 | 8734141 | |||
| X12808.1 | - | 641 | ||||