siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039079.2 | NP_001034168.1 | 15 | 2142 | ||
| NM_008860.3 | NP_032886.2 | 18 | 2342 | |||
| GenBank mRNA | AB110830.1 | - | 15 | 2103 | ||
| AK042562.1 | - | 15 | 2100 | |||
| AK049959.1 | - | 15 | 2126 | |||
| AK147300.1 | - | 15 | 1960 | |||
| BC057694.1 | - | 10 | 1309 | |||
| BC072590.1 | - | 15 | 2142 | |||
| M94632.1 | - | 18 | 2160 | |||

Prkcz
Genomic Map