siRNA Details
| Gene Symbol: Plxna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008882.2 | NP_032908.2 | 14, 15 | 3492 | ||
| GenBank mRNA | AK122289.1 | - | 13, 14 | 1756 | ||
| BC068155.1 | - | 14, 15 | 3274 | 30362726 | ||
| D86949.1 | - | 14, 15 | 3398 | |||

Plxna2
Genomic Map