siRNA Details
| Gene Symbol: Plxna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008883.2 | NP_032909.2 | 3 | 948 | ||
| GenBank mRNA | BC093482.1 | - | 3 | 931 | 30945372 | |
| D86950.1 | - | 3 | 914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxna3| Gene Symbol: Plxna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008883.2 | NP_032909.2 | 3 | 948 | ||
| GenBank mRNA | BC093482.1 | - | 3 | 931 | 30945372 | |
| D86950.1 | - | 3 | 914 | |||