siRNA Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008891.2 | NP_032917.2 | 6 | 522 | ||
| GenBank mRNA | AK009055.1 | - | 6 | 477 | ||
| AK028219.1 | - | 6 | 463 | |||
| AK030978.1 | - | 6 | 451 | |||
| AK050669.1 | - | 6 | 468 | |||
| AK051640.1 | - | 6 | 520 | |||
| AK085295.1 | - | 4 | 971 | |||
| AK157587.1 | - | 6 | 466 | |||
| AK160616.1 | - | 6 | 457 | |||
| AK161109.1 | - | 6 | 462 | |||
| BC132571.1 | - | 6 | 456 | 40130848 | ||
| BC138014.1 | - | 6 | 456 | 8861033 | ||
| Y08701.1 | - | 6 | 453 | |||

Pnn
Genomic Map