siRNA Details
| Gene Symbol: Pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008894.2 | NP_032920.2 | 4 | 497 | ||
| GenBank mRNA | AK013901.1 | - | 4 | 464 | ||
| AK181645.1 | - | 51 | ||||
| AK218431.1 | - | 3 | 212 | |||
| BC003342.1 | - | 4 | 463 | 3257610 | ||
| BG078511.2 | - | 4 | 450 | |||
| CB205181.1 | - | 1 | 33 | |||
| Z72486.1 | - | 4 | 458 | |||

Pold2
Genomic Map